Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (CDP 571 (Humicade)), Alexa Fluor™ 350, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275908AF350
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
TNF-alpha | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TNF | |
Human TNF-alpha | |
0.1 mL | |
Primary | |
The antibody binds to human TNF-alpha with a Kd of 0.087 nM. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry | |
CDP 571 (Humicade) | |
Western Blot, ELISA, Immunohistochemistry | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Human | |
Protein A purified | |
Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
7124.0 | |
Human | |
IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction