Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (D2E7 (Adalimumab)), Alexa Fluor™ 594, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP262567AF594
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.Specifications
TNF-alpha | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence | |
D2E7 (Adalimumab) | |
ELISA, Immunohistochemistry, Immunofluorescence | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha by guided phage-display technology, using the murine anti-hTNF alpha antibody Mab32 as a template (c.f. also US Patent 6090382). | |
0.1 mL | |
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
7124 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction