Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (D2E7 (Adalimumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$269.00
Specifications
Antigen | TNF-alpha |
---|---|
Clone | D2E7 (Adalimumab) |
Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, Block/Neutralize |
Applications | ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization |
Classification | Monoclonal |
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize.Specifications
TNF-alpha | |
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, Block/Neutralize | |
Monoclonal | |
Purified | |
PBS | |
7124 | |
This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha by guided phage-display technology, using the murine anti-hTNF alpha antibody Mab32 as a template (c.f. also US Patent 6090382). | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
D2E7 (Adalimumab) | |
ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization | |
Unconjugated | |
Rabbit | |
APC1 protein, biosimilar, biosimilars, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG κ | |
Binds to soluble TNF-alpha, but not to lymphotoxin (TNF beta). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title