Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (D2E7 (Adalimumab)), PerCP, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP262567PCP
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| TNF-alpha | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Neutralization | |
| D2E7 (Adalimumab) | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Block/Neutralize | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha by guided phage-display technology, using the murine anti-hTNF alpha antibody Mab32 as a template (c.f. also US Patent 6090382). | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction