Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (Hu7), Alexa Fluor™ 488, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB9677G
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Neutralization, Flow CytometrySpecifications
TNF-alpha | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Neutralization, Flow Cytometry | |
Hu7 | |
Neutralization, Intracellular Staining by Flow Cytometry | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Human TNF-alpha | |
0.1 mL | |
Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
7124 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction