Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (J1D9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$631.00
Specifications
Antigen | TNF-alpha |
---|---|
Clone | J1D9 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247676A
![]() |
Novus Biologicals
NBP2476760.1MG |
0.1 mg |
Each for $631.00
|
|
|||||
NBP247676B
![]() |
Novus Biologicals
NBP2476760.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human, Mouse (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, CyTOF-ready.Specifications
TNF-alpha | |
1.0 mg/mL | |
Flow Cytometry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Asthma, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
PBS with No Preservative | |
7124 | |
Recombinant human TNF-alpha | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
17 kDa |
J1D9 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative) | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG1 κ | |
Protein A or G purified | |
This antibody neutralizes TNF alpha biological activities. It prevents TNF alpha induced apoptosis in Jurkat cells. It also neutralizes HurTNFamediated cytotoxicity of L929 cells and inhibits tumor growth in mice. It protects mice against toxicity of HurTNFa. Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia. TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17kDa band following SDS PAGE under non-reducing conditions. TNF alpha is closely related to the 25kDa protein Tumor Necrosis Factor beta (lymphotoxin), sharing the same receptors and cellular actions. TNF alpha causes cytolysis of certain transformed cells, being synergistic with interferon gamma in its cytotoxicity. Although it has little effect on many cultured normal human cells, TNF alpha appears to be directly toxic to vascular endothelial cells. Other actions of TNF alpha include stimulating growth of human fibroblasts and other cell lines, activating polymorphonuclear neutrophils and osteoclasts, and induction of interleukin 1, prostaglandin E2 and collagenase production. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title