Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Antibody (TNF/1500R), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$278.72 - $579.29

Specifications

Antigen TNF-alpha
Clone TNF/1500R
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 4 - 8 ug/ml
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25324220
SDP
View Documents
Novus Biologicals
NBP25324220UG
20 μg
Each for $278.72
Only null left
Add to Cart
 
NB253242100
SDP
View Documents
Novus Biologicals
NBP253242100UG
100 μg
Each for $579.29
Only null left
Add to Cart
 
NB253242200
SDP
View Documents
Novus Biologicals
NBP253242200UG
200 μg N/A N/A N/A
Description

Description

TNF-alpha Monoclonal specifically detects TNF-alpha in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TNF-alpha
0.2 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
7124
Recombinant full-length human TNF-alpha protein
Primary
Store at 4C.
TNF/1500R
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 4 - 8 ug/ml
Monoclonal
Purified
RUO
Human, Rat
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
TNF
IgG κ
Protein A or G purified
17 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.