Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (Z74P2C5*A12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $426.00
Specifications
Antigen | TNF-alpha |
---|---|
Clone | Z74P2C5*A12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
TNF-alpha | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Fish | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG | |
Protein A purified |
Z74P2C5*A12 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
Monoclonal | |
Purified | |
Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
PBS with 0.02% Sodium Azide | |
7124 | |
Ovalbumin-conjugated synthetic peptide | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title