Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNF alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. AMC3012
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMC3012 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMC3012 Supplier Invitrogen™ Supplier No. AMC3012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute using 0.5 mL sterile distilled water.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF alpha
Applications ELISA, Neutralization, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 125mM trehalose and no preservative
Gene Tnf
Gene Accession No. P01375, P06804
Gene Alias APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Gene Symbols Tnf
Host Species Rabbit
Immunogen Recombinant mouse TNF-alpha produced in E. coli.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21926, 7124
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype Ig
Show More Show Less
Which antibodies directed to cytokines are suitable for use in immunohistochemistry with paraffin sections?

The following antibodies have been used successfully for IHC with paraffin sections:
Mouse (monoclonal) Anti-Human TGF-?1 (Cat. No. AHG0051), Rat (monoclonal) Anti-Human Interleukin-13 (Cat. No. AHC0132), Rabbit (polyclonal) Anti-Human IL-8 (Cat. No. AHC0881), Rabbit (polyclonal) Anti-Mouse Interferon-? (Cat. No. AMC4034), Goat (polyclonal) Anti-Mouse TNF-? (Cat. No. AMC3012), and Rabbit (polyclonal) Anti-Rat TNF-? (Cat. No. ARC3012).

You can also locate these and other antibodies with our helpful Primary Antibody Selection Guide. This can be accessed by searching "Primary Antibody Selector Tool" on our website. Once you've reached the Selection Guide, you can narrow the search by clicking on your desired links at the top of the Guide.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.