Learn More
Invitrogen™ TNF alpha Polyclonal Antibody, Biotin
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PBOTNFABI
Description
PBOTNFABI targets TNF Alpha in ELISA, and WB applications and shows reactivity with bovine and ovine samples. The PBOTNFABI immunogen is tNF alpha. PBOTNFABI detects TNF Alpha which has a predicted molecular weight of approximately 26 kDa. The PBOTNFABI polyclonal biotinylated bovine TNFa antibody has successfully been paired as the detection antibody in a sandwich ELISA with monoclonal capture antibody MBOTNFAI (clone E10011). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection.
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
Specifications
TNF alpha | |
Polyclonal | |
Biotin | |
Tnf | |
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
Rabbit | |
Protein G | |
RUO | |
280943, 443540 | |
4°C | |
Lyophilized |
ELISA, Western Blot | |
0.25 mg/mL | |
PBS with Proprietary preservative | |
P23383, Q06599 | |
Tnf | |
TNF alpha. | |
250 μg | |
Primary | |
Bovine, Sheep | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.