Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Rabbit anti-Mouse, Clone: 23, Novus Biologicals™

Rabbit Monoclonal Antibody
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | 23 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Mouse samples. It is validated for Sandwich ELISA Capture.Specifications
| TNF-alpha | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Recombinant Mouse TNF-alpha Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 23 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 7124 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title