Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF, anti-Human, Clone: CDP 571 (Humicade), Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00089
Specifications
| TNF | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| DIF/TNF-alpha/TNFA/TNFSF2 | |
| Original antibody is raised against native purified human TNF. | |
| RUO | |
| 7124 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA | |
| CDP 571 (Humicade) | |
| ELISA (0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| tumor necrosis factor (TNF superfamily, member 2) | |
| TNF | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction