Learn More
Invitrogen™ TNF beta Monoclonal Antibody (LTX-21), FITC, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS105FI

Description
Description: TNF-beta (LTX-21) is a mouse monoclonal antibody raised against native and recombinant TNF-beta of human origin.BMS105 specifically recognizes natural TNF-beta as well as recombinant TNF-beta (16 kDa and 18 kDa form). The antibody can be used to stain TNF-beta producing cells. Immunoperoxidase or immunofluorescence techniques are applicable. BMS105 is also suitable as primary antibody in staining for FACS analysis,for for Western blot and ELISA application. BMS105FI can be used for direct staining of cells. Applications Tested: Flow Cytomtery. Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
Specifications
TNF beta | |
Monoclonal | |
10 μL/Test | |
TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 | |
P01374 | |
LTA | |
Affinity Chromatography | |
RUO | |
4049 | |
4°C, store in dark | |
Liquid |
Flow Cytometry | |
LTX-21 | |
FITC | |
LTA | |
DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF ß; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFß; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.