Learn More
TNF Mouse, anti-Human, R718, Clone: MAB11, BD Horizon™
Mouse Monoclonal Antibody
Supplier:  BD Biosciences 566957
 View more versions of this product
Catalog No. BDB566957
Only null left
Add to Cart
Description
Description
The MAb11 monoclonal antibody specifically binds to human tumor necrosis factor (TNF, also known as TNF-α) protein. TNF is an efficient juxtacrine, paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The MAb11 antibody has been reported to crossreact with Rhesus Macaque TNF.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNF
Monoclonal
R718
100 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
MAB11
Mouse
RUO
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.