Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF, Mouse Anti-Human, RY610, Clone: MAb11, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 571327
Description
TNF Monoclonal antibody specifically detects TNF in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
TNF | |
Monoclonal | |
RY610 | |
Tumor necrosis factor alpha; TNF-a; TNF-α; TNFSF2; Cachectin | |
Recombinant Human TNF | |
25 Tests | |
Primary | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
Purified |
Flow Cytometry (Intracellular Staining) | |
MAb11 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
7124 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction