Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ TNF (Mouse) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
Description
TNF (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse TNF alpha in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).
Specifications
Specifications
| Cross Reactivity | Mouse |
| For Use With (Application) | Quantitative |
| Gene | Tnf |
| Gene Alias | DIF, MGC151434, TNF-alpha, TNFSF2, TNFalpha, Tnfa, Tnfsf1a |
| Gene ID (Entrez) | 21926 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction