Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-α Rat anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: MP6-XT3, Southern Biotech™
SDP

Catalog No. OB1022914 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1022914 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1022914 Supplier Southern Biotechnology Associates Supplier No. 1022914
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen TNF-α
Applications ELISA, Dot Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Low Endotoxin/Azide-free (LE/AF)
Clone MP6-XT3
Concentration 0.5mg/mL
Formulation Supplied in PBS
Host Species Rat
Immunogen E. coli-expressed mouse TNF-α
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20°C
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.