Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF RI/TNFRSF1A Antibody (16803R), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB225RW
Description
TNF RI/TNFRSF1A Monoclonal antibody specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Flow Cytometry, NeutralizationSpecifications
TNF RI/TNFRSF1A | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Neutralization | |
16803R | |
Flow Cytometry, Neutralization | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
E. coli-derived recombinant human TNF RI/TNFRSF1A, Accession # NP_001056 | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Signal Transduction | |
7132 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction