Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF RI/TNFRSF1A Antibody (RM0011-6F13), Alexa Fluor™ 700

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB11085469AF700
Description
TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
TNF RI/TNFRSF1A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFRSF1A | |
Mouse TNF RI/TNFRSF1A (extracellular domain) | |
0.1 mL | |
Apoptosis, Cancer, Cytokine Research, Phospho Specific | |
7132 | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
RM0011-6F13 | |
Western Blot, Flow Cytometry, ELISA | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
This is specific to the extracellular domain of mouse TNFR-1 protein. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction