Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF RII/TNFRSF1B Antibody (001), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290948H
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Cynomolgus Monkey samples. It is validated for ELISA.Specifications
TNF RII/TNFRSF1B | |
Monoclonal | |
HRP | |
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus TNF RII/TNFRSF1B (F7EAE4; Met1-Asp257). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
7133 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction