Learn More
TNF RII/TNFRSF1B Antibody (RM0012-6F13) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | TNF RII/TNFRSF1B |
| Applications | CyTOF, Flow Cytometry, ELISA, Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | RM0012-6F13 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:100-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:100-1:200, CyTOF-ready |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Gene Alias | CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II |
| Gene Symbols | TNFRSF1B |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.