Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF RII/TNFRSF1B Antibody (RM0012-6F13), Biotin, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB11085470B
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, ELISA.Specifications
TNF RII/TNFRSF1B | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TNFRSF1B | |
Mouse TNF RII/TNFRSF1B (extracellular domain) | |
0.1 mL | |
Primary | |
This is specific to the extracellular domain of mouse TNFR-II protein. | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA | |
RM0012-6F13 | |
Western Blot, Flow Cytometry, ELISA | |
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
Armenian Hamster | |
Protein A or G purified | |
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
7133 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction