Learn More
TNFAIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 153201AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0147
Only null left
Add to Cart
Description
Description
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.
Specifications
Specifications
TNFAIP1
Polyclonal
Unconjugated
TNFAIP1
B12, B61, BACURD2, EDP1, hBACURD2, Protein B12, TNFAIP1
Rabbit
Antigen Affinity Chromatography
RUO
21927, 287543, 7126
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13829, Q7TNY1
Tnfaip1
TNFAIP1 Fusion Protein Ag0220
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.