Learn More
Invitrogen™ TNFAIP3 Recombinant Rabbit Monoclonal Antibody (0K8B0)

Description
Immunogen sequence: PEESTGGPHS APPTAPSPFL FSETTAMKCR SPGCPFTLNV QHNGFCERCH NARQLHASHA PDHTRHLDPG KCQACLQDVT RTFNGICSTC FKRTTAEASS S.
Specifications
Specifications
| Antigen | TNFAIP3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 0K8B0 |
| Concentration | 0.95 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TNFAIP3 |
| Gene Accession No. | P21580 |
| Gene Alias | A20; A20p37; A20p50; AISBL; OTU domain-containing protein 7C; OTUD7C; Putative DNA-binding protein A20; RP11-356I2.3; TNF alpha induced protein 3; TNF alpha-induced protein 3; TNFA1P2; TNFAIP3; TNFAIP3 (A20); Tnfip3; Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; zinc finger protein A20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.