Learn More
Tnfaip6 Polyclonal Antibody, Invitrogen™
Description
Immunogen sequence: DDPGCLADYV EIYDSYDDVH GFVGRYCGDE LPDDIISTGN VMTLKFLSDA SVTAGGFQIK YVAMDPVSKS SQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Specifications
Specifications
| Antigen | Tnfaip6 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TNFAIP6 |
| Gene Accession No. | P98066 |
| Gene Alias | Hyaluronate-binding protein; TNF alpha induced protein 6; TNF alpha-induced protein 6; Tnfaip6; Tnfip6; TNF-stimulated gene 6 protein; TSG6; TSG-6; tumor necrosis factor alpha induced protein 6; tumor necrosis factor alpha-induced protein 6; tumor necrosis factor alpha-inducible protein 6; tumor necrosis factor induced protein 6; tumor necrosis factor, alpha induced protein 6; tumor necrosis factor, alpha-induced protein 6; tumor necrosis factor-inducible gene 6 protein; tumor necrosis factor-stimulated gene-6 protein |
| Gene Symbols | TNFAIP6 |
| Show More |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.