Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFAIP8 Antibody (SR1270), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TNFAIP8 |
---|---|
Clone | SR1270 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
TNFAIP8 Monoclonal antibody specifically detects TNFAIP8 in Human samples. It is validated for Western BlotSpecifications
TNFAIP8 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GG2-1, Head and neck tumor and metastasis-related protein, MDC-3.13TNF-induced protein GG2-1, NDED, NF-kappa-B-inducible DED-containing protein, SCC-S2SCCS2, TNF alpha-induced protein 8, tumor necrosis factor alpha-induced protein 8, tumor necrosis factor, alpha-induced protein 8 | |
A synthesized peptide derived from human TNFAIP8 (Uniprot #: O95379) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1270 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
25816 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title