Learn More
TNFAIP8 Mouse anti-Human, Clone: OTI1H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810682

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as a negative mediator of apoptosis and may play a role in tumor progression. Suppresses the TNF-mediated apoptosis by inhibiting caspase-8 activity but not the processing of procaspase-8, subsequently resulting in inhibition of BID cleavage and caspase-3 activation.Specifications
TNFAIP8 | |
Monoclonal | |
Unconjugated | |
TNFAIP8 | |
GG2-1, MDC-3.13, NDED, SCC-S2, SCCS2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
25816 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95379 | |
TNFAIP8 | |
Full length human recombit protein of human TNFAIP8 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.