Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFAIP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563397 Supplier Invitrogen™ Supplier No. PA563397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKSHGRVNNV FDHFSDCEFL AALYNPFGNF KPHLQKLCDG INKMLDEENI Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

TNFalpha-IP 8 (tumor necrosis factor, alpha-induced protein 8), also known as NFkappa B-inducible DED-containing protein (NDED), SCC-S2 or TNF-induced protein GG2-1, is a 198 amino acid cytoplasmic protein induced by NFkappa B and TNF. The induction of TNFalpha-IP 8 by TNF is dependent on the activation of NFkappa B. TNFalpha-IP 8 negatively mediates apoptosis and may also play a role in tumor progression. TNFalpha-IP 8 specifically inhibits caspase-8 activity, which results in the inhibition of BID cleavage and caspase-3 activation during TNF-mediated apoptosis. TNFalpha-IP 8 is expressed at high levels in thymus, bone marrow, lymph node, spleen, thyroid, placenta and various tumor tissues, as well as fetal lung, liver and kidney. TNFalpha-IP 8 is present as three isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tnfaip8
Gene Accession No. O95379
Gene Alias AA987150; E130304C20Rik; ENSMUSG00000073567; GG2-1; Gm10539; head and neck tumor and metastasis-related protein; MDC3.13; MDC-3.13; Nded; NF-kappa-B-inducible DED-containing protein; SCC S2; SCCS2; SCC-S2; Ssc-2; TNF alpha induced protein 8; TNF alpha-induced protein 8; Tnfaip8; TNFalpha IP8; TNFalpha-IP 8; TNFalphaIP8; TNF-induced protein; TNF-induced protein GG2-1; Tumor necrosis factor alpha-induced protein 8; tumor necrosis factor, alpha induced protein 8; tumor necrosis factor, alpha-induced protein 8
Gene Symbols Tnfaip8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNFAIP8. Recombinant protein control fragment (Product #RP-101818).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25816
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.