Learn More
Invitrogen™ TNFAIP8 Recombinant Rabbit Monoclonal Antibody (24GB2315)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557785

Description
TNFAIP8 Recombinant Monoclonal Antibody for Western Blot, Flow
TNFalpha-IP 8 (tumor necrosis factor, alpha-induced protein 8), also known as NFkappa B-inducible DED-containing protein (NDED), SCC-S2 or TNF-induced protein GG2-1, is a 198 amino acid cytoplasmic protein induced by NFkappa B and TNF. The induction of TNFalpha-IP 8 by TNF is dependent on the activation of NFkappa B. TNFalpha-IP 8 negatively mediates apoptosis and may also play a role in tumor progression. TNFalpha-IP 8 specifically inhibits caspase-8 activity, which results in the inhibition of BID cleavage and caspase-3 activation during TNF-mediated apoptosis. TNFalpha-IP 8 is expressed at high levels in thymus, bone marrow, lymph node, spleen, thyroid, placenta and various tumor tissues, as well as fetal lung, liver and kidney. TNFalpha-IP 8 is present as three isoforms produced by alternative splicing.
Specifications
TNFAIP8 | |
Recombinant Monoclonal | |
Unconjugated | |
Tnfaip8 | |
AA987150; E130304C20Rik; ENSMUSG00000073567; GG2-1; Gm10539; head and neck tumor and metastasis-related protein; MDC3.13; MDC-3.13; Nded; NF-kappa-B-inducible DED-containing protein; SCC S2; SCCS2; SCC-S2; Ssc-2; TNF alpha induced protein 8; TNF alpha-induced protein 8; Tnfaip8; TNFalpha IP8; TNFalpha-IP 8; TNFalphaIP8; TNF-induced protein; TNF-induced protein GG2-1; Tumor necrosis factor alpha-induced protein 8; tumor necrosis factor, alpha induced protein 8; tumor necrosis factor, alpha-induced protein 8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
106869, 25816, 307428 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
24GB2315 | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95379, Q921Z5 | |
Tnfaip8 | |
A synthesized peptide derived from human TNFAIP8 (aa 1-35). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.