Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFAIP8L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569889 Supplier Invitrogen™ Supplier No. PA569889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 100%.

Tumor necrosis factor, α-induced protein 8-like 1, also known as TNFα-IP 8L1, is a 186 amino acid protein belonging to the TNFα-IP 8 family. Members of the TNFα-IP 8 family are induced by nuclear factor-κB 9 (NFκB) and tumor necrosis factor (TNF), although induction by TNF is dependent upon NFκB activation. TNFα-IP 8 proteins also act as a negative mediator of apoptosis and may play a role in tumor progression. They suppress TNF-mediated apoptosis by inhibiting caspase-8 activity but not the processing of procaspase-8, subsequently resulting in inhibition of BID cleavage and caspase-3 activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP8L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TNFAIP8L1
Gene Accession No. Q8WVP5
Gene Alias 2600017J23Rik; Oxidative stress regulated gene-beta; oxidative stress-regulated gene-beta; Oxy-beta; TIPE1; TNF alpha induced protein 8 like 1; TNF alpha-induced protein 8-like protein 1; Tnfaip8l1; TNFAIP8-like protein 1; Tumor necrosis factor alpha-induced protein 8-like protein 1; tumor necrosis factor, alpha induced protein 8 like 1; tumor necrosis factor, alpha-induced protein 8-like 1; tumor necrosis factor, alpha-induced protein 8-like protein 1
Gene Symbols TNFAIP8L1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TNFAIP8L1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126282
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.