Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNFAIP8L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111745
Description
Immunogen sequence: PAARSMDSDS GEQSEGEPVT AAGPDV.
Acts as a lipid transfer protein. Preferentially captures and shuttles two lipid second messengers, i.e., phosphatidylinositol 4,5- bisphosphate and phosphatidylinositol 3,4,5-trisphosphate and increases their levels in the plasma membrane. Additionally, may also function as a lipid-presenting protein to enhance the activity of the PI3K-AKT and MEK-ERK pathways. May act as a regulator of tumorigenesis through its activation of phospholipid signaling. [UniProt].
Specifications
| TNFAIP8L3 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP8L3 | |
| TIPE3; TNF alpha induced protein 8 like 3; TNF alpha-induced protein 8-like protein 3; TNFAIP8L3; TNFAIP8-like protein 3; Tumor necrosis factor alpha-induced protein 8-like protein 3; tumor necrosis factor, alpha induced protein 8 like 3; tumor necrosis factor, alpha-induced protein 8-like 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5GJ75 | |
| TNFAIP8L3 | |
| Recombinant Protein corresonding to Human TNFAIP8L3. Recombinant protein control fragment (Product #RP-108683). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction