Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676412
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-412 Supplier Invitrogen™ Supplier No. PA140293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-40293 detects TNF-R I from human samples. PA1-40293 has been successfully used in ELISA, functional assays, immunoprecipitation and Western blot applications. The PA1-40293 imunogen is the extracellular region of mouse TNF-R I. Endotoxin <24 EU/mg.

TNFR1 (Tumor necrosis factor receptor 1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. TNFR1 plays an important role in mediating, in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of TNFR1 is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12. Anti-apoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with TNFR1, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of TNFR1 were found to be associated with the autosomal dominant periodic fever syndrome, and the impaired receptor clearance is thought to be a mechanism of the disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFR1
Applications ELISA, Functional Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Tnfrsf1a
Gene Accession No. P19438
Gene Alias CD120a; CD120a antigen; FPF; hypothetical protein; metalloproteinase inhibitor 4; MGC19588; MS5; n-smase activation assoc. factor; p55; p55 TNF receptor; p55-R; p60; p62; p63; sCD120a; soluble CD120a; soluble TNFR1; sTNF RI; sTNFR I; sTNFR1; sTNFRI; TBP1; TBPI; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4; TNF receptor alpha chain; TNF receptor superfamily member 1A; TNF-alphaR1; TNF-alpha-R1; TNFalpha-R1; TNFAR; TNFR; TNF-R; TNFR I; TNFR1; TNF-R1; Tnfr-1; TNFR1-d2; Tnfr-2; TNFR55; TNF-R55; TNFR60; TNFRI; TNF-RI; TNF-R-I; TNFR-I; TNFRp55; Tnfrsf1a; tumor necrosis factor binding protein 1; Tumor necrosis factor receptor; tumor necrosis factor receptor 1; tumor necrosis factor receptor 1A isoform beta; tumor necrosis factor receptor superfamily member 1A; Tumor necrosis factor receptor superfamily member 1A, membrane form; tumor necrosis factor receptor superfamily, member 1A; tumor necrosis factor receptor type 1; tumor necrosis factor receptor type I; tumor necrosis factor-alpha receptor; Tumor necrosis factor-binding protein 1
Gene Symbols Tnfrsf1a
Host Species Rabbit
Immunogen Extracellular region of murine TNF-R I.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7132
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.