Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFR2 Monoclonal Antibody (MR2-1), Biotin, Invitrogen™

Catalog No. FERMA549131
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549131 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549131 Supplier Thermo Scientific Supplier No. MA549131

Mouse Monoclonal Antibody

The antibody reacts with the extra-cellular part of the TNF-RII. It also reacts with the soluble receptor. The antibody cross reacts with rhesus and cynomolgus natural TNF-RII.

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFR2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Inhibition Assays, Western Blot
Classification Monoclonal
Clone MR2-1
Concentration 100 μg/mL
Conjugate Biotin
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Tnfrsf1b
Gene Accession No. P20333
Gene Alias CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Gene Symbols Tnfrsf1b
Host Species Mouse
Immunogen sTNF-R75 purified from the culture supernatant of NSO cells (transfected with the extracellular part of TNF-R75)
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7133
Target Species Human
Content And Storage 4° C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.