Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586383 Supplier Invitrogen™ Supplier No. PA586383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Tnfrsf1b
Gene Accession No. P20333, P25119, Q80WY6
Gene Alias CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Gene Symbols Tnfrsf1b
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 390-440 of Human TNF-R2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 156767, 21938, 7133
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.