Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA517363
Description
It is not recommended to aliquot this antibody.
TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Specifications
TNFR2 | |
Polyclonal | |
TNFRSF1B | |
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
156767, 21938, 7133 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
P20333, P25119, Q80WY6 | |
TNFRSF1B | |
Synthetic peptide corresponding to residues surrounding Asp339 within the intracellular region of human TNF-R2 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction