Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFR2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 50558726 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-558-726 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-558-726 Supplier Proteintech Group Inc Supplier No. 192721AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.

Specifications

Antigen TNFR2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNFRSF1B
Gene Accession No. P20333, P25119
Gene Alias CD120b, Etanercept, p75, p75TNFR, p80 TNF alpha receptor, TBP 2, TBPII, TNF R II, TNF R2, TNF R75, TNF Receptor 2, TNF Receptor II, TNF RII, TNFBR, TNFR II, TNFR1B, TNFR2, TNFR80, TNFRSF1B
Gene Symbols TNFRSF1B
Host Species Rabbit
Immunogen TNFR2 Fusion Protein Ag5866
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21938, 7133
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.