Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNFR2 (soluble) Monoclonal Antibody (KT30)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA172521
Description
TNFR2 (soluble) Monoclonal Antibody for ELISA
TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Specifications
TNFR2 (soluble) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P20333 | |
Tnfrsf1b | |
Recombinant soluble human TNFRII. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA | |
KT30 | |
Unconjugated | |
Tnfrsf1b | |
CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2 | |
Mouse | |
Protein A | |
RUO | |
7133 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction