Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFRH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547723
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547723 Supplier Invitrogen™ Supplier No. PA547723
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TNF RI and rmTNF RII is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The TNFRH3 gene encodes a protein that is part of the tumor necrosis factor receptor superfamily, which is involved in regulating immune responses, inflammation, and apoptosis. TNFRH3, also identified as tumor necrosis factor receptor homolog 3, is characterized by its similarities to other TNF receptors, yet its specific ligand and signaling pathways remain an area of active research. TNF receptors typically engage in critical interactions with ligands such as TNF-α, facilitating cellular responses that include the modulation of immune cell function, survival, and proliferation. Although TNFRH3's precise function is not fully elucidated, it is hypothesized to play a role in modulating immune signaling and inflammatory responses, potentially impacting autoimmune disease processes and cancer progression by influencing apoptosis and cell survival. Like other members of the TNF receptor family, TNFRH3 might interact with adaptor proteins to form signaling complexes that propagate downstream effects critical for immune homeostasis. Research efforts continue to define its role in physiological and pathological states, aiming to leverage its function for therapeutic applications in treating inflammatory diseases and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRH3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Tnfrsf26
Gene Accession No. P83626
Gene Alias TNF receptor homolog 3; TNFR; Tnfrh3; Tnfrsf24; Tnfrsf26; Tumor necrosis factor receptor; Tumor necrosis factor receptor superfamily member 26; tumor necrosis factor receptor superfamily, member 24; tumor necrosis factor receptor superfamily, member 26
Gene Symbols Tnfrsf26
Host Species Sheep
Immunogen CHO-derived recombinant mouse TNFRH3/TNFRSF26 Ser20-Lys162.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244237
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.