Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRH3/TNFRSF26 Antibody (509027), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5330F
Description
TNFRH3/TNFRSF26 Monoclonal antibody specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
TNFRH3/TNFRSF26 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8718 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
509027 | |
Flow Cytometry, CyTOF-ready | |
APO-3;DDR3;DR3;GEF720;LARD;PLEKHG5;TNFRSF12;TNFRSF25;TR3;TRAMP;Tumor necrosis factor receptor superfamily member 25;WSL-1;WSL-LR | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26, Ser20-Lys162, Accession # P83626 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction