Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNFRSF11B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA519841
Description
For Western Blot, this antibody has non-specific bands at 105 kDa and 14 kDa. This antibody is predicted to react with mouse, sheep, cow and pig based on sequence homology.
The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
Specifications
TNFRSF11B | |
Polyclonal | |
Unconjugated | |
TNFRSF11B | |
LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25341, 4982 | |
-20°C or -80°C if preferred | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.9 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
O00300, O08727 | |
TNFRSF11B | |
Synthetic peptide conjugated to KLH derived from within residues 200 - 300 Osteoprotegerin. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction