Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFRSF11B Recombinant Rabbit Monoclonal Antibody (340)
GREENER_CHOICE

Catalog No. PIMA530947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530947 Supplier Invitrogen™ Supplier No. MA530947
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Osteoprotegerin/TNFRSF11B.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRSF11B
Applications ELISA
Classification Recombinant Monoclonal
Clone 340
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNFRSF11B
Gene Accession No. O00300
Gene Alias LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
Gene Symbols TNFRSF11B
Host Species Rabbit
Immunogen Recombinant Human Osteoprotegerin/TNFRSF11B protein (Met1-Leu401).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.