Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRSF13B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TNFRSF13B |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3461
![]() ![]() |
Proteintech Group Inc
215201AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Simultaneously four different laboratories identified a a new member of the tumor necrosis factor (TNF) family. This has been named as TALL-1 (1), THANK (TNF homologue that activates apoptosis, nuclear factor-kappaB, and c-Jun NH2-terminal kinase (2), BAFF (for B cell activating factor belonging to the TNF family) (3) and BlyS (B lymphocyte stimulator). Membrane-bound BAFF is processed and secreted through the action of a protease whose specificity matches that of the furin family of proprotein convertases. The receptor for BlyS/BAFF appears to be expressed predomitly on B cells and activated T cells. Recently two orphan receptors TACI (6) and BCMA (,8) have been shown to be receptors for BlyS/BAFF (6-9). TACI is expressed on B cells and signals through CAML, activating the transcription factors NF-AT, NF-kB, and AP-1. TACI is a type III transmembrane protein, as defined by the lack of a cleaved signal sequence and its N-terminal extracellular exposure. The cDNA codes for 293 amino acids with predicted molecular weight of approximately 32 kDa.Specifications
TNFRSF13B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O14836 | |
23495 | |
TNFRSF13B Fusion Protein Ag16097 | |
Primary | |
-20°C | |
TNFRSF13B |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD267, CVID, TACI, TNFRSF13B, TNFRSF14B | |
TNFRSF13B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title