Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFRSF14 Recombinant Rabbit Monoclonal Antibody (004)
GREENER_CHOICE

Catalog No. PIMA535989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535989 Supplier Invitrogen™ Supplier No. MA535989
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

TNFRSF14 Recombinant Monoclonal Antibody for Flow

TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRSF14
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 004
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNFRSF14
Gene Accession No. Q92956
Gene Alias Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509
Gene Symbols TNFRSF14
Host Species Rabbit
Immunogen Recombinant Human HVEM/TNFRSF14/CD270 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8764
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.