Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF18 (soluble form) Mouse anti-Human, Clone: T10P2H9*, B9, MilliporeSigma™

Catalog No. MABF952MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF952MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF952MI Supplier MilliporeSigma Supplier No. MABF952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-TNFRSF18 (soluble form) Antibody, clone T10P2H9&asterisk;B9 is tested in Immunohistochemistry (Paraffin) and Western Blotting for the detection of TNFRSF18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRSF18 (soluble form)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T10P2H9*B9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TNFRSF18,AITR,GITR,UNQ319/PRO364
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to 9 amino acids from isoform 2 of human TNFRSF18.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_004186
Test Specificity Clone T10P2H9*B9 specifically detects the soluble form of TNFRSF18 in MCF-7 cells. It recognizes an epitope of 9 amino acids in the C-terminal region of isoform 2 of TNFRSF18.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.