Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF19 Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
SDP

Catalog No. 501683331
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3331 Supplier OriGene Supplier No. TA810325
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.

Specifications

Antigen TNFRSF19
Applications Western Blot
Classification Monoclonal
Clone OTI1B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TNFRSF19
Gene Accession No. Q9NS68
Gene Alias TAJ, TAJ-alpha, TRADE, TROY
Gene Symbols TNFRSF19
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 30-170 of human TNFRSF19 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55504
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.