Learn More
Invitrogen™ TNFRSF19L Monoclonal Antibody (3F8)

Description
Peptide Sequence: STTLWQCPPG EEPDLDPGQG TLCRPCPPGT FSAAWGSSPC QPHARCSLWR RLEAQVGMAT RDTLCGDCWP GWFGPWGVPR VPCQPCSWAP LGTHGCDEW.
Specifications
Specifications
| Antigen | TNFRSF19L |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3F8 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RELT |
| Gene Accession No. | Q969Z4 |
| Gene Alias | E430021K24Rik; Receptor expressed in lymphoid tissues; Relt; RELT tumor necrosis factor receptor; Tnfrsf19l; TRLT; Tumor necrosis factor receptor superfamily member 19L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.