Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNFRSF19L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112840
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112840 Supplier Invitrogen™ Supplier No. PA5112840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TNFRSF19L Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is especially abundant in hematologic tissues. It has been shown to activate the NF-kappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1). This receptor is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRSF19L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RELT
Gene Accession No. Q969Z4
Gene Alias E430021K24Rik; Receptor expressed in lymphoid tissues; Relt; RELT tumor necrosis factor receptor; Tnfrsf19l; TRLT; Tumor necrosis factor receptor superfamily member 19L
Gene Symbols RELT
Host Species Rabbit
Immunogen Recombinant Human Tumor necrosis factor receptor superfamily member 19L protein (26-153AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84957
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.