Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRSF4 Mouse anti-Human, Clone:8H6H7, Abnova™
Mouse monoclonal antibody raised against partial recombinant human TNFRSF4.
Supplier: Abnova Corporation MAB17973
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that this receptor promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation. [provided by RefSeq]Specifications
TNFRSF4 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
TNFRSF4 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
8H6H7 | |
tumor necrosis factor receptor superfamily, member 4 | |
In PBS (0.05% sodium azide) | |
ACT35, CD134, OX40, TXGP1L | |
Recombinant protein corresponding to amino acids 29-214 of human TNFRSF4. | |
7293 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction