Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF4 Mouse anti-Human, Clone: OTI2F12A2, liquid, TrueMAB™
SDP

Catalog No. 501684532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4532 Supplier OriGene Supplier No. TA812359
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OX40 (CD134) is a member of the TNF-receptor superfamily, and has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that OX40 promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation.OX40 is not constitutively expressed on resting naive T cells, unlike CD28. OX40 is a secondary costimulatory molecule, expressed after 24 to 72 hours following activation. OX40L, a ligand of OX40, is also not expressed on resting antigen presenting cells. Expression of OX40 is dependent on full activation of the T cell; without CD28, expression of OX40 is delayed and expressed at lower levels.

Specifications

Antigen TNFRSF4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2F12A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TNFRSF4
Gene Accession No. P43489
Gene Alias ACT35, CD134, IMD16, OX40, TXGP1L
Gene Symbols TNFRSF4
Host Species Mouse
Immunogen Full length human recombit protein of human TNFRSF4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7293
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.