Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF8, anti-Human, Clone: Ki-4, Abnova™

Catalog No. 89032369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-369 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-369 Supplier Abnova Corporation Supplier No. RAB00026
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen TNFRSF8
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone Ki-4
Conjugate Unconjugated
Dilution ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene tumor necrosis factor receptor superfamily, member 8
Gene Alias CD30/D1S166E/KI-1
Gene Symbols TNFRSF8
Immunogen Original antibody is raised against native purified human TNFRSF8.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Test Specificity This antibody binds to TNFRSF8.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.